Learn More
Cayman Chemical Lipid U-101, Purity: ≥95%, CAS: 3049129-09-8, 50 mg
SDP
Catalog No. 502978651
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid U-101 and encapsulating mRNA encoding a fusion protein composed of IL-2, a linker, and CD25 inhibit tumor growth in an MC-38 mouse xenograft model
Catalog No. 50-297-8651 Supplier Cayman Chemical Supplier No. 4090250 MG
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid U-101 and encapsulating mRNA encoding a fusion protein composed of IL-2, a linker, and CD25 inhibit tumor growth in an MC-38 mouse xenograft model