Learn More
PRECIGENOME LipidFlex™ RNA-LNP SM-102 kit, LNP Formulation for RNA/DNA encapsulation
SDP
Catalog No. NC3781969
Encompass
LipidFlex™ RNA-LNP kit is a 4-component lipid mixture. This highly efficient lipid formulation is used to synthesize mRNA lipid nanoparticles for non-viral gene delivery. Using NanoGenerator Flex-S system and mixer cartridge, customers can prepare potent mRNA LNP in a convenient and efficient way.Narrow size distribution of mRNA LNP· High transfection efficiency· High protein expression level· High cell viability· Time efficient synthesis process
Catalog No. NC3781969 Supplier PRECIGENOME Supplier No. PGLPFRNA02Q125UL
LipidFlex™ RNA-LNP kit is a 4-component lipid mixture. This highly efficient lipid formulation is used to synthesize mRNA lipid nanoparticles for non-viral gene delivery. Using NanoGenerator Flex-S system and mixer cartridge, customers can prepare potent mRNA LNP in a convenient and efficient way.Narrow size distribution of mRNA LNP· High transfection efficiency· High protein expression level· High cell viability· Time efficient synthesis process