Learn More
Cayman Chemical LIpIdLaunchALC0315 LNP KIt 1ea
SDP
Catalog No. 502895772
Encompass_Preferred
Cayman’s LipidLaunch™ ALC-0315 LNP Kit (Loadable) provides cargo-ready, empty ALC-0315 LNPs. These LNPs are capable of rapid encapsulation of nucleic acids, without the need for incubation steps, enabling subsequent delivery to target cells or other downstream research applications. LipidLaunch™ LNPs (Loadable) demonstrate very low toxicity compared to traditional lipid-based transfection methods. Conventionally loaded LNPs typically require specialized equipment to ensure consistent control of particle size, while also requiring large reaction volumes. LipidLaunch™ ALC-0315 LNPs (Loadable) enable users to encapsulate cargo in low reaction volumes offering greater flexibility in the scale of preparation while preserving costly nucleic acid cargo.
Catalog No. 50-289-5772 Supplier Cayman Chemical Supplier No. 7027501 EA
Add to Cart
Add to Cart
Cayman’s LipidLaunch™ ALC-0315 LNP Kit (Loadable) provides cargo-ready, empty ALC-0315 LNPs. These LNPs are capable of rapid encapsulation of nucleic acids, without the need for incubation steps, enabling subsequent delivery to target cells or other downstream research applications. LipidLaunch™ LNPs (Loadable) demonstrate very low toxicity compared to traditional lipid-based transfection methods. Conventionally loaded LNPs typically require specialized equipment to ensure consistent control of particle size, while also requiring large reaction volumes. LipidLaunch™ ALC-0315 LNPs (Loadable) enable users to encapsulate cargo in low reaction volumes offering greater flexibility in the scale of preparation while preserving costly nucleic acid cargo.