Learn More
Cayman Chemical LipidLaunch™ LNP-0315 Uptake Kit (Green Fluorescence), 1 ea
SDP
Catalog No. 502916440
Encompass_Preferred
Cayman’s LipidLaunch™ LNP-0315 Uptake Kit (Green Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs by their target cell type without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-291-6440 Supplier Cayman Chemical Supplier No. 382191 EA
Cayman’s LipidLaunch™ LNP-0315 Uptake Kit (Green Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs by their target cell type without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.