Learn More
Cayman Chemical LipidLaunch™ LNP-0315 Uptake Kit (Near-Infrared Fluorescence), 1 ea
SDP
Catalog No. 502916126
Encompass_Preferred
Cayman’s LipidLaunch™ LNP-0315 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-291-6126 Supplier Cayman Chemical Supplier No. 390641 EA
Cayman’s LipidLaunch™ LNP-0315 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.