Learn More
Cayman Chemical LIpIdLaunchLNP102 Uptake K 1ea
SDP
Catalog No. 502917229
Encompass_Preferred
Cayman’s LipidLaunch™ LNP-102 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-291-7229 Supplier Cayman Chemical Supplier No. 390651 EA
Cayman’s LipidLaunch™ LNP-102 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.