Learn More
Cayman Chemical LIpIdLaunchLNPMC3 Uptake K 1ea
SDP
Catalog No. 502918657
Encompass_Preferred
Cayman’s LipidLaunch™ LNP-MC3 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-291-8657 Supplier Cayman Chemical Supplier No. 390661 EA
Cayman’s LipidLaunch™ LNP-MC3 Uptake Kit (Near-Infrared Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs in vivo without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.