Learn More
Cayman Chemical LIpIdLaunchLNPMC3 Uptake K 1ea
SDP
Catalog No. 502917257
Encompass_Preferred
Cayman’s LipidLaunch™ LNP-MC3 Uptake Kit (Green Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs by their target cell type without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-291-7257 Supplier Cayman Chemical Supplier No. 390671 EA
Add to Cart
Add to Cart
Cayman’s LipidLaunch™ LNP-MC3 Uptake Kit (Green Fluorescence) is intended to serve as a starting point for laboratories to explore the uptake of LNPs by their target cell type without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.