Learn More
Cayman Chemical LipidLaunch™ SORT LNP Exploration Kit, 1 ea
SDP
Catalog No. 502942217
Encompass_Preferred
Cayman’s LipidLaunch™ SORT LNP Exploration Kit is intended to serve as a starting point for laboratories to explore the feasibility of using SORT LNPs for their individual application without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.
Catalog No. 50-294-2217 Supplier Cayman Chemical Supplier No. 400991 EA
Cayman’s LipidLaunch™ SORT LNP Exploration Kit is intended to serve as a starting point for laboratories to explore the feasibility of using SORT LNPs for their individual application without the need for specialized equipment. Optimal preparation conditions for the encapsulation of nucleic acids with LNPs must be determined by the end user.