Learn More
Biotium LipidSpot 488, fluorogenic neutral lipid stain, green fluorescence, 1000X concentration, DMSO solvent, 2 to 8 °C storage
Catalog No. NC1701657
Encompass_Preferred
LipidSpot 488, fluorogenic neutral lipid stain, green fluorescence, 1000X concentration, DMSO solvent, 2 to 8 °C storage
Catalog No. NC1701657 Supplier Biotium Supplier No. 70065
LipidSpot 488, fluorogenic neutral lipid stain, green fluorescence, 1000X concentration, DMSO solvent, 2 to 8 °C storage