Learn More
Biotium LipidSpot™ Lipid Droplet Stains 610, 125ul
Catalog No. NC1701659
Encompass_Preferred
LipidSpot™ stains are fluorescent dyes that rapidly stain lipid droplets in live or fixed cells, with no wash step and minimal background.Rapidly and specifically stain lipid droplets. Minimal background, no wash required.
Catalog No. NC1701659 Supplier Biotium Supplier No. 70069
LipidSpot™ stains are fluorescent dyes that rapidly stain lipid droplets in live or fixed cells, with no wash step and minimal background.Rapidly and specifically stain lipid droplets. Minimal background, no wash required.