Learn More
Aviva Systems Biology Corp LIPJ Antibody - C-terminal region (ARP51815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIPJ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LIPJ. Alternate names: FLJ11218, LIPL1, bA425M17.2ApplicationWB. Species reactivity: Cow, Horse, Human, Rabbit, Rat
Catalog No. 50-014-60623