Learn More
Aviva Systems Biology Corp LIPM Antibody - N-terminal region (ARP64348_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64348_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIPM. It was validated on Western Blot. . Alternate names: LIPL3, bA304I5.1ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-288-242