Learn More
Medchemexpress LLC Lipoamide-PEG3-mal | 1314378-19-2 | 99.6% | C25H41N3O7S2 | 10MG
SDP
Catalog No. 5000261319
Encompass_Preferred
Lipoamide-PEG3-Mal is a PEG-based linker that combines a lipoamide (lipoic acid derivative) moiety with a terminal maleimide. It is supplied for research use as a building block in the synthesis of PROTACs and other bioconjugates, providing flexible spacing and a thiol-reactive handle for selective conjugation.
  • Peg3 spacer provides flexibility and improved solubility.
  • Lipoamide moiety offers a bio-relevant functional group for linkage.
  • Maleimide group enables rapid and selective thiol conjugation.
  • High reported purity suitable for synthetic applications.
  • Available in small, research-friendly package sizes.
Catalog No. 50-002-61319 Supplier Medchemexpress LLC Supplier No. HY14133810MG
Lipoamide-PEG3-Mal is a PEG-based linker that combines a lipoamide (lipoic acid derivative) moiety with a terminal maleimide. It is supplied for research use as a building block in the synthesis of PROTACs and other bioconjugates, providing flexible spacing and a thiol-reactive handle for selective conjugation.
  • Peg3 spacer provides flexibility and improved solubility.
  • Lipoamide moiety offers a bio-relevant functional group for linkage.
  • Maleimide group enables rapid and selective thiol conjugation.
  • High reported purity suitable for synthetic applications.
  • Available in small, research-friendly package sizes.