Learn More
Medchemexpress LLC Lipoamido-PEG8-acid | 1334172-70-1 | MFCD21363243 | 95% | C27H51NO11S2 | 25 MG
SDP
Catalog No. 5000352903
Encompass_Preferred
Lipoamido-PEG8-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It contains a lipoic acid group and a carboxylic acid moiety. The hydrophilic PEG linker increases the water solubility of the compound. Storage conditions: -20°C.
  • Contains a lipoic acid group and a carboxylic acid moiety
  • Terminal carboxylic acid reacts with primary amine groups
  • Hydrophilic PEG linker increases water solubility
Catalog No. 50-003-52903 Supplier Medchemexpress LLC Supplier No. 25MGHY141333
Add to Cart
Add to Cart
Lipoamido-PEG8-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It contains a lipoic acid group and a carboxylic acid moiety. The hydrophilic PEG linker increases the water solubility of the compound. Storage conditions: -20°C.
  • Contains a lipoic acid group and a carboxylic acid moiety
  • Terminal carboxylic acid reacts with primary amine groups
  • Hydrophilic PEG linker increases water solubility