Learn More
Medchemexpress LLC Lipocalin-2/NGAL Cy 10ug | 10ug
SDP
Catalog No. 5000194805
Encompass_Preferred
Lipocalin-2/NGAL is a multifaceted iron transporter involved in multiple biological processes including apoptosis innate immunity and kidney development Lipocalin-2 dynamically affects cellular iron concentration through binding to the siderophore 2 3-dihydroxybenzoate Lipocalin-2/NGAL Protein Cynomolgus (CHO His) is the recombinant cynomolgus-derived Lipocalin-2/NGAL protein expressed by CHO with C-His labeled tag
Catalog No. 50-001-94805 Supplier Medchemexpress LLC Supplier No. HYP7911410UG
Lipocalin-2/NGAL is a multifaceted iron transporter involved in multiple biological processes including apoptosis innate immunity and kidney development Lipocalin-2 dynamically affects cellular iron concentration through binding to the siderophore 2 3-dihydroxybenzoate Lipocalin-2/NGAL Protein Cynomolgus (CHO His) is the recombinant cynomolgus-derived Lipocalin-2/NGAL protein expressed by CHO with C-His labeled tag