Learn More
AdipoGen Lipocalin-2 rat pAb
SDP
Supplier:  AdipoGen AG25A0088C100
Encompass_Preferred
Polyclonal Antibody. Recognizes rat lipocalin-2. Detects a band of ~25kDa by Western blot. Cross-reacts with human and mouse lipocalin-2. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Lipocalin-2 is an iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development.
Catalog No. 50-256-3231