Learn More
AdipoGen Lipocalin-2 rat rec
SDP
Catalog No. 502562996
Encompass_Preferred
Protein. Rat lipocalin-2 aa 1-198 is fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Lipocalin-2 is an iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development.
Catalog No. 50-256-2996 Supplier AdipoGen Supplier No. AG40A0108C010
Protein. Rat lipocalin-2 aa 1-198 is fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Lipocalin-2 is an iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development.