Learn More
Sino Biological Lipocalin-2/LCN2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10222MM18100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Lipocalin-2/LCN2 (rh Lipocalin-2/LCN2; Catalog#10222-H08H; NP_005555.2; Met1-Gly198). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8143