Learn More
SignaGen Laboratories LipoD293™ In Vitro DNA Transfection Reagent 1.0mL
Catalog No. 504782
Encompass
Features- Top choice for hard-to-transfect cells- Exceptional high titers of virus production- Equally good for very long DNAs (up to 180 kb)- Equally good for suspension 293 cells (e.g., 293F, 293H, etc)- High levels of recombinant protein production- Presence of serum and antibiotics enhances efficiency on 293 cells- Exceptional efficiency for both single DNA transfection and multi DNAs co-transfection- Very affordableStorage ConditionStore at 4 °C. If stored properly, the product is stable for 12 months or longer.
Catalog No. 50-478-2 Supplier SignaGen Laboratories Supplier No. SL100668
Features- Top choice for hard-to-transfect cells- Exceptional high titers of virus production- Equally good for very long DNAs (up to 180 kb)- Equally good for suspension 293 cells (e.g., 293F, 293H, etc)- High levels of recombinant protein production- Presence of serum and antibiotics enhances efficiency on 293 cells- Exceptional efficiency for both single DNA transfection and multi DNAs co-transfection- Very affordableStorage ConditionStore at 4 °C. If stored properly, the product is stable for 12 months or longer.