Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
GREENER_CHOICE

Catalog No. 11668500
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.3 mL
0.75 mL
1.5 mL
15 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-668-500 15 mL
11-668-019 1.5 mL
11-668-027 0.75 mL
11-668-030 0.3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-668-500 Supplier Invitrogen™ Supplier No. 11668500
Only null left
Add to Cart
Edge
Add to Cart
Request a Quote

Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines

Lipofectamine™ 2000 Transfection Reagent is versatile transfection reagent that has been shown to effectively transfect widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine 2000 Transfection Reagent is best transfection reagent available from Life Technologies for the co-transfection of siRNA and plasmid DNA.

  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum, eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines

Order Info

Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one bottle (15 mL) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 15 mL
Serum Compatible Yes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.