Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Catalog No. CMAX00003
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.1 mL
0.3 mL
0.75 mL
1.5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CMAX00003 0.3 mL
CMAX00008 0.75 mL
CMAX00001 0.1 mL
CMAX00015 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CMAX00003 Supplier Invitrogen™ Supplier No. CMAX00003
  • $316.00
    $293.65 / Each of 1

    Save $22.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is an optimized lipid-based transfection reagent for CRISPR-Cas9 protein delivery, providing the cleavage efficiency of electroporation with simplicity and scalability.

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid-based transfection reagent for CRISPR knockout applications. It effectively delivers the Cas9 protein and guide RNA (gRNA) ribonucleoprotein (RNP) complex, ensuring high transfection efficiency and effective gene knockout. The proprietary lipid-based transfection reagent minimizes cellular toxicity, making it suitable to for sensitive cells. Its ease of use and compatibility with high-throughput screening make it a cost-effective alternative to electroporation.

Combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it provides:

  • Optimized for CRISPR—specially formulated for CRISPR-Cas9 RNP applications
  • Superior gene knockout—efficient Cas9 transfection ensures higher genomic cleavage
  • Broad cell type applicability—versatile for use in a wide range of cell lines, including primary cells and stem cells
  • Minimal cytotoxicity—viable, healthy cells maintained throughout Cas9 transfection
  • High-throughput compatible—an ideal delivery solution for functional genomic studies with CRISPR-Cas9

Achieve high transfection efficiency in novel genome editing applications

Lipofectamine CRISPRMAX Transfection Reagent enables successful Cas9 transfection in a wide range of cell types, including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and more. Unlike CRISPR plasmid or Cas9 mRNA, this protein-based approach eliminates the need for transcription or translation, reducing the risk of genomic integration and making the process cell cycle-independent. When combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it ensures superior cleavage efficiency and effective gene editing.

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Protein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Product Line CRISPRMAX, Lipofectamine
Product Type Transfection Reagent
Quantity 0.3 mL
Serum Compatible Yes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less
Can I use Lipofectamine CRISPRMAX Cas9 Transfection Reagent with one-cell embryos?

We have no data regarding the use of CRISPRMAX in one-celled embryos, therefore we do not recommend using this product for this application.

The shipping for my Lipofectamine CRISPRMAX Cas9 Transfection Reagent was delayed. Can I still use it?

All of the Lipofectamine reagents, including Lipofectamine CRISPRMAX Cas9 Transfection Reagent, are stable at room temperature for months. What is most important is that the reagent did not freeze, because that will alter the integrity and composition of the lipid particles and will compromise usability. Previous freezing can be evidenced by precipitation and excessive cloudiness of the reagent. A limited amount of cloudiness is normal, but excessive cloudiness is a sign that the reagent froze at some point. If the reagent you received looks clear or a bit cloudy the product should still be usable. Some additional data on room temperature stability of some Lipofectamine reagents, in the context of shipping, can be found in this white paper.

How much Lipofectamine CRISPRMAX Reagent, Cas9 protein, and gRNA should I complex for gene editing?

Please see the Lipofectamine CRISPRMAX Reagent manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0014545_lipofectamine_crispermax_QR.pdf) for protocol details or contact techsupport@thermofisher.com.

How does the Lipofectamine CRISPRMAX Reagent work?

The Lipofectamine CRISPRMAX Reagent combined with the proprietary enhancement properties of the Lipofectamine Cas9 Plus Reagent leads to efficient complex formation with the Cas9-gRNA ribonucleoprotein, for best delivery to the nucleus, helping to ensure high gene editing frequency for a wide range of cell types.

Can the Lipofectamine CRISPRMAX Reagent be used to deliver proteins other than the Cas9 nuclease?

Although the Lipofectamine CRISPRMAX Reagent was developed for the delivery of the Cas9-gRNA ribonucleoprotein, there is potential for other protein complex applications.

Can the Lipofectamine CRISPRMAX Reagent also be used to deliver the donor plasmid in addition to the Cas9 nuclease protein and gRNA?

The Lipofectamine CRISPRMAX Reagent was developed to efficiently deliver the Cas9-gRNA ribonucleoprotein complex. We recommend that you first deliver the donor plasmid with Lipofectamine 3000, then follow with delivering the Cas9-gRNA complex with Lipofectamine CRISPRMAX Reagent for best editing efficiency.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.