Learn More
Medchemexpress LLC Lipofermata | 250MG
SDP
Catalog No. 5000308182
Encompass_Preferred
Lipofermata is a fatty acid transport protein 2 (FATP2) inhibitor Lipofermata shows fatty acid transport inhibition with an IC50 of 4 84 M in Caco-2 cells Lipofermata an analog of spiro-indoline-thadiazole shows zinc-specific suppression of antibacterial activity Lipofermata perturbs zinc homeostasis in E coli K-12 with a MIC of 16 M[1][2][3]
Catalog No. 50-003-08182 Supplier Medchemexpress LLC Supplier No. HY116788250MG
Lipofermata is a fatty acid transport protein 2 (FATP2) inhibitor Lipofermata shows fatty acid transport inhibition with an IC50 of 4 84 M in Caco-2 cells Lipofermata an analog of spiro-indoline-thadiazole shows zinc-specific suppression of antibacterial activity Lipofermata perturbs zinc homeostasis in E coli K-12 with a MIC of 16 M[1][2][3]