Learn More
TARGETMOL CHEMICALS INC LIPOFERMATA 5MG
SDP
Catalog No. 502577441
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lipofermata is a fatty acid transport protein (FATP) inhibitor that prevents lipid transport to melanoma cells and reduces melanoma growth and invasion. purity: 99%
Catalog No. 50-257-7441 Supplier TARGETMOL CHEMICALS INC Supplier No. T118545MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lipofermata is a fatty acid transport protein (FATP) inhibitor that prevents lipid transport to melanoma cells and reduces melanoma growth and invasion. purity: 99%