Learn More
Medchemexpress LLC α-Lipoic Acid | 1077-28-7 | MFCD00005441 | 206.33 | 1 G
SDP
Catalog No. 5000390081
Encompass_Preferred
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. This product can also be used to construct self-assembled nanoaggregates with improved antitumor efficacy.
  • Antioxidant properties
  • Essential cofactor for mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis
  • Supports research in infection, inflammation, and cancer
Catalog No. 50-003-90081 Supplier Medchemexpress LLC Supplier No. HYN04921G
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. This product can also be used to construct self-assembled nanoaggregates with improved antitumor efficacy.
  • Antioxidant properties
  • Essential cofactor for mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis
  • Supports research in infection, inflammation, and cancer