Learn More
Medchemexpress LLC α-Lipoic Acid | 1077-28-7 | MFCD00005474 | 206.33 | 5 G
SDP
Catalog No. 5000389715
Encompass_Preferred
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used with CPUL1 to construct the self-assembled nanoaggregate CPUL1-LA NA, which demonstrates improved antitumor efficacy compared to CPUL1.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used to construct self-assembled nanoaggregates with improved antitumor efficacy
Catalog No. 50-003-89715 Supplier Medchemexpress LLC Supplier No. HYN04925G
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used with CPUL1 to construct the self-assembled nanoaggregate CPUL1-LA NA, which demonstrates improved antitumor efficacy compared to CPUL1.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used to construct self-assembled nanoaggregates with improved antitumor efficacy