Learn More
Medchemexpress LLC Lipoic acid | 1200-22-2 | 968259 | 99.9% | 206.33 | 5 G
SDP
Catalog No. 5000385723
Encompass_Preferred
Lipoic acid ((R)-(+)-α-Lipoic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • More effective than racemic lipoic acid
  • Biosynthesized in humans and essential for aerobic metabolism
  • Nutritionally and therapeutically preferred form due to its "vitamin-like" role in metabolism
  • Exists in nature
Catalog No. 50-003-85723 Supplier Medchemexpress LLC Supplier No. HY187335G
Lipoic acid ((R)-(+)-α-Lipoic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • More effective than racemic lipoic acid
  • Biosynthesized in humans and essential for aerobic metabolism
  • Nutritionally and therapeutically preferred form due to its "vitamin-like" role in metabolism
  • Exists in nature