Learn More
Medchemexpress LLC HY-N0492 500mg Medchemexpress, α-Lipoic Acid CAS:1077-28-7 Purity:>98%
SDP
Catalog No. 501970279
Encompass_Preferred
Medchemexpress, HY-N0492 500mg α-Lipoic Acid CAS:1077-28-7 α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0279 Supplier Medchemexpress LLC Supplier No. HYN0492500MG
Medchemexpress, HY-N0492 500mg α-Lipoic Acid CAS:1077-28-7 α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.