Learn More
Medchemexpress LLC α-Lipoic acid sodium | 2319-84-8 | 228.31 | 1 G
SDP
Catalog No. 5000390204
Encompass_Preferred
α-Lipoic Acid (Thioctic acid) sodium is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation. It also induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells and can be used with CPUL1 to form self-assembled nanoaggregate CPUL1-LA NA, which shows improved antitumor efficacy.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used with CPUL1 to form self-assembled nanoaggregate CPUL1-LA NA
Catalog No. 50-003-90204 Supplier Medchemexpress LLC Supplier No. HYN0492A1G
α-Lipoic Acid (Thioctic acid) sodium is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation. It also induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells and can be used with CPUL1 to form self-assembled nanoaggregate CPUL1-LA NA, which shows improved antitumor efficacy.
  • Antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used with CPUL1 to form self-assembled nanoaggregate CPUL1-LA NA