Learn More
Medchemexpress LLC Α-Lipoic acid sodium | 2319-84-8 | 228.31 | 5 G
SDP
Catalog No. 5000389349
Encompass_Preferred
α-Lipoic Acid sodium is an antioxidant that serves as an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
  • Serves as an antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Supports formation of self-assembled nanoaggregates for improved antitumor efficacy
Catalog No. 50-003-89349 Supplier Medchemexpress LLC Supplier No. HYN0492A5G
α-Lipoic Acid sodium is an antioxidant that serves as an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
  • Serves as an antioxidant
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Supports formation of self-assembled nanoaggregates for improved antitumor efficacy