Learn More
Medchemexpress LLC α-Lipoic Acid sodium (Thioctic acid sodium) | 2319-84-8 | 228.31 | 500 MG
SDP
Catalog No. 5000389223
Encompass_Preferred
α-Lipoic Acid sodium (Thioctic acid sodium) is a potent antioxidant that acts as an essential cofactor for mitochondrial enzyme complexes. It plays a role in cellular processes, inhibiting NF-κB-dependent HIV-1 LTR activation and inducing endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
  • Potent antioxidant properties
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used to construct self-assembled nanoaggregates for improved antitumor efficacy
Catalog No. 50-003-89223 Supplier Medchemexpress LLC Supplier No. HYN0492A500MG
α-Lipoic Acid sodium (Thioctic acid sodium) is a potent antioxidant that acts as an essential cofactor for mitochondrial enzyme complexes. It plays a role in cellular processes, inhibiting NF-κB-dependent HIV-1 LTR activation and inducing endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
  • Potent antioxidant properties
  • Essential cofactor of mitochondrial enzyme complexes
  • Inhibits NF-κB-dependent HIV-1 LTR activation
  • Induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells
  • Can be used to construct self-assembled nanoaggregates for improved antitumor efficacy