Learn More
Medchemexpress LLC Α-lipoic Acid (Standard) | 1077-28-7 | 206.33 | 50 MG
SDP
Catalog No. 5000390115
Encompass_Preferred
α-Lipoic Acid (Standard) is the analytical standard of α-Lipoic Acid, intended for research and analytical applications. As an antioxidant and essential cofactor of mitochondrial enzyme complexes, α-Lipoic Acid (Thioctic acid) inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used to construct the self-assembled nanoaggregate CPUL1-LA NA, which has improved antitumor efficacy.
  • The compound is the grade of analytical standard.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-90115 Supplier Medchemexpress LLC Supplier No. HYN0492R50MG
α-Lipoic Acid (Standard) is the analytical standard of α-Lipoic Acid, intended for research and analytical applications. As an antioxidant and essential cofactor of mitochondrial enzyme complexes, α-Lipoic Acid (Thioctic acid) inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used to construct the self-assembled nanoaggregate CPUL1-LA NA, which has improved antitumor efficacy.
  • The compound is the grade of analytical standard.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.