Learn More
Medchemexpress LLC Lipoic acid (Standard) | 1200-22-2 | 99.8% | 206.33 | 100 MG
SDP
Catalog No. 5000387872
Encompass_Preferred
Lipoic acid (Standard) is the analytical standard of Lipoic acid, intended for research and analytical applications. It acts as an antioxidant and is an essential cofactor of mitochondrial enzyme complexes. The (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid. This compound is used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Acts as an antioxidant
  • Serves as an essential cofactor of mitochondrial enzyme complexes
  • More effective than racemic Lipoic acid
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
Catalog No. 50-003-87872 Supplier Medchemexpress LLC Supplier No. HY18733R100MG
Lipoic acid (Standard) is the analytical standard of Lipoic acid, intended for research and analytical applications. It acts as an antioxidant and is an essential cofactor of mitochondrial enzyme complexes. The (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid. This compound is used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Acts as an antioxidant
  • Serves as an essential cofactor of mitochondrial enzyme complexes
  • More effective than racemic Lipoic acid
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS