Learn More
Medchemexpress LLC α-Lipoic Acid (Thioctic acid) | 1077-28-7 | MFCD00005474 | 206.33 | 1 ML
SDP
Catalog No. 5000389714
Encompass_Preferred
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used with CPUL1 (HY-151802) to form self-assembled nanoaggregate CPUL1-LA NA, which shows improved antitumor efficacy compared to CPUL1. This product is for research use only.
  • Plays an essential role in mitochondrial bioenergetics.
  • Reduces lipid accumulation in the liver.
  • Increases NAD+/NADH ratio in HepG2 cells.
  • Increases SIRT1 activity in HepG2 cells.
  • Increases phosphorylation of AMPK and acetyl-CoA carboxylase (ACC) in HepG2 cells.
  • Markedly reduces visceral fat mass in C57BL/6J mice.
  • Inhibits appetite and causes significant weight loss in mice.
Catalog No. 50-003-89714 Supplier Medchemexpress LLC Supplier No. HYN049210MM/1ML
α-Lipoic Acid (Thioctic acid) is an antioxidant and an essential cofactor of mitochondrial enzyme complexes. It inhibits NF-κB-dependent HIV-1 LTR activation and induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells. It can also be used with CPUL1 (HY-151802) to form self-assembled nanoaggregate CPUL1-LA NA, which shows improved antitumor efficacy compared to CPUL1. This product is for research use only.
  • Plays an essential role in mitochondrial bioenergetics.
  • Reduces lipid accumulation in the liver.
  • Increases NAD+/NADH ratio in HepG2 cells.
  • Increases SIRT1 activity in HepG2 cells.
  • Increases phosphorylation of AMPK and acetyl-CoA carboxylase (ACC) in HepG2 cells.
  • Markedly reduces visceral fat mass in C57BL/6J mice.
  • Inhibits appetite and causes significant weight loss in mice.