Learn More
LIPOID LLC LIPOID PE 18:0/18:0 PEG 2000, PEGylated Phospholipid, 2000 Da, >99% purity, sodium salt, -20°C storage, soluble in ethanol at 5 mg/mL
Catalog No. NC3397654
Encompass
LIPOID PE 18:0/18:0 PEG 2000, PEGylated Phospholipid, 2000 Da, >99% purity, sodium salt, -20°C storage, soluble in ethanol at 5 mg/mL
Catalog No. NC3397654 Supplier LIPOID LLC Supplier No. 588209RS1
LIPOID PE 18:0/18:0 PEG 2000, PEGylated Phospholipid, 2000 Da, >99% purity, sodium salt, -20°C storage, soluble in ethanol at 5 mg/mL