Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharide from Po1MG
Catalog No. 502904509
Encompass
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4) a member of the Toll-like receptor protein family which recognizes common pathogen-associated molecular-patterns (PAMPs).
Catalog No. 50-290-4509 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SMB006101MG
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4) a member of the Toll-like receptor protein family which recognizes common pathogen-associated molecular-patterns (PAMPs).