Learn More
Biomatik Corporation Lipopolysaccharide (Lps) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30370
Encompass_Preferred
Lipopolysaccharide (Lps) Monoclonal Antibody is a Universal Monoclonal Antibody to Lipopolysaccharide (LPS). The Lipopolysaccharide (Lps) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Universal. Applications: WB, IHC. Target Synonyms: Lipopolysaccharide (LPS). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5818