Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Lipopolysaccharide (LPS) Solution (500X)

Catalog No. 501122025
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2025 100 μL
50-112-9325 4 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2025 Supplier Invitrogen™ Supplier No. 00497693
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Intended for in vitro culture of B cells, monocytes, macrophages, and other cells that are responsive to LPS.

  • A ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution
  • LPS is a major component of the cell wall of gram negative bacteria
  • It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex
  • This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS
TRUSTED_SUSTAINABILITY

Specifications

Cell Type B cells, monocytes, macrophages and other cells that are responsive to LPS.
Concentration 500 X
Form Liquid
Product Type LPS Solution
Sterility Sterile
Product Line eBioscience
Quantity 100 μL
Can I freeze thaw eBioscience Lipopolysaccharide (LPS) Solution (500X)?

eBioscience Lipopolysaccharide (LPS) Solution (500X) has been tested under conditions where no change in performance was observed after 20 freeze-thaw cycles. For optimal performance, we recommend preparing smaller aliquots to minimize the number of freeze-thaw cycles.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.