Learn More
ACROBiosystems Recombinant Protein;Human LBP /Lipopolysaccharide Protein;HEK293;1MG;LBP,Lipopolysaccharide-binding protein,LBP,LPS
Supplier:  ACROBiosystems LBPH52H51MG
Encompass_Preferred
Recombinant Protein;Human LBP /Lipopolysaccharide Protein;1MG;Human Lipopolysaccharide-binding Protein ,His Tag, Ala 26 - Val 481 (Accession # P18428), was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.8 kDa. The protein migrates as 62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8835