Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from Escherichia coli O111:B4, 297-473-0, MFCD00164401, 10 x 1mg
Catalog No. NC1615973
Encompass
γ-irradiated, BioXtra, suitable for cell culture, Synonym: LPS; This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.
Catalog No. NC1615973 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L439110X1MG
γ-irradiated, BioXtra, suitable for cell culture, Synonym: LPS; This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.