Learn More
Selleck Chemical LLC Lipopolysaccharides
SDP
Catalog No. 5000003591
Encompass_Preferred
Lipopolysaccharides (LPSs Endotoxin LPS) is an endotoxin derived from the outer leaflet of the outer membrane of Gram-negative bacteria Lipopolysaccharides consists of an antigen O-specific chain a core oligosaccharide and lipid A Lipopolysaccharides is a pathogenic associated molecular pattern (PAMP) that activates the immune system Lipopolysaccharides activates TLR-4 on immune cells Lipopolysaccharides induces secretion of cell migrasome
Catalog No. 50-000-03591 Supplier Selleck Chemical LLC Supplier No. S78505MG
Lipopolysaccharides (LPSs Endotoxin LPS) is an endotoxin derived from the outer leaflet of the outer membrane of Gram-negative bacteria Lipopolysaccharides consists of an antigen O-specific chain a core oligosaccharide and lipid A Lipopolysaccharides is a pathogenic associated molecular pattern (PAMP) that activates the immune system Lipopolysaccharides activates TLR-4 on immune cells Lipopolysaccharides induces secretion of cell migrasome