Learn More
Medchemexpress LLC HY-D1056 10mg , Lipopolysaccharides CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD105610MG
Encompass_Preferred
HY-D1056 10mg Lipopolysaccharides CAS: Lipopolysaccharides (LPS) is an endotoxin derived from the outer leaflet of the outer membrane of Gram-negative bacteria. Lipopolysaccharides consists of an antigen O-specific chain, a core oligosaccharide and lipid A. Lipopolysaccharides is a pathogenic associated molecular pattern (PAMP) that activates the immune system. Lipopolysaccharides activates TLR-4 on immune cells. This product is derived from Escherichia coli O55:B5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1162