Learn More
Medchemexpress LLC Lipopolysaccharides, from E. coli O26:B6 | 10-100 kDa | 10 MG
SDP
Catalog No. 5000297176
Encompass_Preferred
Lipopolysaccharides from Escherichia coli O26:B6 are purified endotoxins used to activate Toll-like receptor 4 and stimulate innate immune responses in vitro and in vivo. They are commonly used to model inflammation, sepsis, and acute lung injury and for testing immune-modulating compounds.
  • Purified endotoxin that stimulates TLR4 signaling.
  • Suitable for in vitro and in vivo inflammation models.
  • White to off-white solid; soluble in water (≈50 mg/mL with sonication).
  • Store sealed away from moisture; in solvent store -80°C (6 months) or -20°C (1 month).
  • Documentation available: datasheet, COA, SDS, handling instructions.
Catalog No. 50-002-97176 Supplier Medchemexpress LLC Supplier No. HYD1056A310MG
Lipopolysaccharides from Escherichia coli O26:B6 are purified endotoxins used to activate Toll-like receptor 4 and stimulate innate immune responses in vitro and in vivo. They are commonly used to model inflammation, sepsis, and acute lung injury and for testing immune-modulating compounds.
  • Purified endotoxin that stimulates TLR4 signaling.
  • Suitable for in vitro and in vivo inflammation models.
  • White to off-white solid; soluble in water (≈50 mg/mL with sonication).
  • Store sealed away from moisture; in solvent store -80°C (6 months) or -20°C (1 month).
  • Documentation available: datasheet, COA, SDS, handling instructions.