Learn More
Medchemexpress LLC Lipopolysaccharides, from E. coli O55:B5 (LPS) | 93572-42-0 | MFCD00164401 | 100 MG
SDP
Catalog No. 5000428268
Encompass_Preferred
Lipopolysaccharides, from E. coli O55:B5 (LPS, from Escherichia coli (O55:B5)) are endotoxins and TLR4 activators extracted from Escherichia coli (E. coli O55:B5) and are classified as S (smooth) type LPS. They possess the typical three-part structure: O-antigen, core oligosaccharide, and lipid A. Lipopolysaccharides, from E. coli O55:B5 activate TLR-4 in immune cells, exhibit high pyrogenicity, and demonstrate dose and serotype specificity. They can be widely used to induce cellular inflammation and establish animal models related to inflammation.
  • Endotoxins and TLR4 activators
  • Extracted from Escherichia coli (E. coli O55:B5)
  • Classified as S (smooth) type LPS
  • Possess typical three-part structure: O-antigen, core oligosaccharide, and lipid A
  • Activate TLR-4 in immune cells
  • Exhibit high pyrogenicity
  • Demonstrate dose and serotype specificity
Catalog No. 50-004-28268 Supplier Medchemexpress LLC Supplier No. HYD1056100MG
Lipopolysaccharides, from E. coli O55:B5 (LPS, from Escherichia coli (O55:B5)) are endotoxins and TLR4 activators extracted from Escherichia coli (E. coli O55:B5) and are classified as S (smooth) type LPS. They possess the typical three-part structure: O-antigen, core oligosaccharide, and lipid A. Lipopolysaccharides, from E. coli O55:B5 activate TLR-4 in immune cells, exhibit high pyrogenicity, and demonstrate dose and serotype specificity. They can be widely used to induce cellular inflammation and establish animal models related to inflammation.
  • Endotoxins and TLR4 activators
  • Extracted from Escherichia coli (E. coli O55:B5)
  • Classified as S (smooth) type LPS
  • Possess typical three-part structure: O-antigen, core oligosaccharide, and lipid A
  • Activate TLR-4 in immune cells
  • Exhibit high pyrogenicity
  • Demonstrate dose and serotype specificity