Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides E100MG
Catalog No. 502904416
Encompass
This product is phenol extracted from E. coli serotype O127B8. The source strain is ATCC 12740. This LPS serotype has been used in the study of septic shock and to induce NOS in murine macrophages and PAf synthesis in rat glomerular masangial cells.
Catalog No. 50-290-4416 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L3129100MG
Add to Cart
Add to Cart
This product is phenol extracted from E. coli serotype O127B8. The source strain is ATCC 12740. This LPS serotype has been used in the study of septic shock and to induce NOS in murine macrophages and PAf synthesis in rat glomerular masangial cells.