Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides E500MG
Catalog No. 502914653
Encompass
This product is TCA extracted from E. coli serotype O111B4. The source strain is from a private collection. This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.
Catalog No. 50-291-4653 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L4130500MG
This product is TCA extracted from E. coli serotype O111B4. The source strain is from a private collection. This LPS serotype has been used to stimulate B-cells and induce NOS in human hepatocytes.