Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lipopolysaccharides from Escherichia coli O127:B8 | 10 MG
SDP

Catalog No. 5000297687
Encompass_Preferred

Lipopolysaccharides from E. coli O127:B8 are endotoxins extracted from Escherichia coli (O127:B8) that activate Toll-like receptor 4 (TLR-4). They are used to induce inflammatory responses in vitro and in vivo and to construct intestinal inflammation models.

  • Solid, white to off-white appearance for easy visual identification.
  • Soluble in water at ~1 mg/mL with gentle warming and sonication.
  • Heterogeneous molecular weight (typically 10-100 kDa or higher), reflecting natural variability.
  • Storage: solid at 4°C sealed; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available in multiple pack sizes, including 10 mg.
  • Documentation available: data sheet, COA, SDS, and handling instructions.

Catalog No. 50-002-97687 Supplier Medchemexpress LLC Supplier No. HYD1056A210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipopolysaccharides from E. coli O127:B8 are endotoxins extracted from Escherichia coli (O127:B8) that activate Toll-like receptor 4 (TLR-4). They are used to induce inflammatory responses in vitro and in vivo and to construct intestinal inflammation models.

  • Solid, white to off-white appearance for easy visual identification.
  • Soluble in water at ~1 mg/mL with gentle warming and sonication.
  • Heterogeneous molecular weight (typically 10-100 kDa or higher), reflecting natural variability.
  • Storage: solid at 4°C sealed; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available in multiple pack sizes, including 10 mg.
  • Documentation available: data sheet, COA, SDS, and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.