Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L56682ML
Encompass
This product is a 0.2 um filtered aqueous SOLUTIONon containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O127B8. This LPS serotype has been used in studies of septic shock to induce neutrophils to induce NOS in rat brain and induce PAf synthesis in rat glomerular mesangial cells.
Catalog No. 50-290-4933