Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L55432ML
Encompass
This product is a 0.2 um filtered aqueous SOLUTIONon containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O26B6. This LPS serotype has been used in studies of septic shock.
Catalog No. 50-290-4417