Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from E
Catalog No. 502904242
Encompass
This product is a 0.2 um filtered aqueous SOLUTIONon containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O55B5. The source strain is CDC 1644-70. This LPS serotype has been used in studies of septic shock and to induce myocarditis.
Catalog No. 50-290-4242 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L54182ML
This product is a 0.2 um filtered aqueous SOLUTIONon containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O55B5. The source strain is CDC 1644-70. This LPS serotype has been used in studies of septic shock and to induce myocarditis.